Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-p-tolyl ether, 99%

Catalog No. p-7052653
Click to view available options
Quantity:
5 g
25 g

CAS: 1579-40-4 | C14H14O | 198.265 g/mol

Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.

Solubility
Soluble in acetone, benzene, ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1579-40-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00025979
InChI Key YWYHGNUFMPSTTR-UHFFFAOYSA-N
Synonym p-tolyl ether, di-p-tolyl ether, 4-tolyl ether, 4,4'-oxybis methylbenzene, bis 4-methylphenyl ether, 4,4'-dimethyldiphenyl ether, benzene, 1,1'-oxybis 4-methyl, p-p-tolyloxy toluene, 1,1'-oxybis 4-methylbenzene
PubChem CID 74098
IUPAC Name 1-methyl-4-(4-methylphenoxy)benzene
SMILES CC1=CC=C(C=C1)OC2=CC=C(C=C2)C

Specifications

Melting Point 47°C to 49°C
Color Yellow
Boiling Point 285°C
Flash Point >113°C (235°F)
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in acetone,benzene,ether.
Formula Weight 198.26
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Di-p-tolyl ether
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 216-423-0
RTECSNumber DA6625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.