Di-tert-butyl dicarbonate, 97+%

Manufacturer: Alfa Aesar A1470822

Quantity 100g

 View more versions of this product

Catalog No. AAA1470822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24424-99-5
    Molecular Formula C10H18O5
    Formula Weight 218.25
    MDL Number MFCD00008805
    UN Number 2930
    Beilstein 1911173
    Chemical Name or Material Di-tert-butyl Dicarbonate

    Specifications

    Formula Weight 218.25
    Quantity 100g
    UN Number 2930
    Melting Point 21° to 24°C
    Boiling Point 56° to 57°C (0.5mmHg)
    Flash Point 37°C (99°F)
    TSCA TSCA
    MDL Number MFCD00008805
    Beilstein 1911173
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H330-H318-H228-H315-H317-H335
    Fatal if inhaled.
    Causes serious eye damage.
    Flammable solid.
    Causes skin irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P304+P340-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and e
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, FLAMMABLE, ORGANIC, N.O.S.
    RTECS Number HT0230000
    Density 0.950
    Assay Percent Range ≥97%
    CAS 24424-99-5
    Molecular Formula C10H18O5
    Storage Note 1 Moisture sensitive
    Chemical Name or Material Di-tert-butyl Dicarbonate
    Assay ≥97%

    Description

    Applications
    Reagent for the introduction of the Boc protecting group.Di-tert-butyl dicarbonate is a reagent used for the introduction of BOC protecting group. It plays an important role in the preparation of 6-acetyl-1,2,3,4-tetrahydropyridine by reacting with 2-piperidone. It serves as a protecting group used in solid phase peptide synthesis.

    Solubility
    Miscible with decalin, toluene, carbon tetrachloride, tetrahydrofuran, dioxane, alcohols, acetone, acetonitrile and dimethylformamide. Immiscible with water.

    Notes
    Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Moisture and light sensitive. Incompatible with oxidizing agents, reducing agents, bases and acids.

    GHS H Statement
    H330-H318-H228-H315-H317-H335
    Fatal if inhaled.
    Causes serious eye damage.
    Flammable solid.
    Causes skin irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P280-P305+P351+P338-P304+P340-P310-P302+P352
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and e

    Moisture sensitive

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit