Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl dicarbonate, 99%

Catalog No. AC194675000
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC194675000

Supplier: Thermo Scientific Chemicals 194675000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 24424-99-5 | C10H18O5 | 218.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Melting Point 22.0°C to 24.0°C
Color White
Density 1.0200g/mL
Boiling Point 56.0°C to 57.0°C (0.5 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4075 to 1.4095
Linear Formula O[CO2C(CH3)3]2
Quantity 500 g
Fieser 04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
Specific Gravity 1.02
Merck Index 15, 3038
Formula Weight 218.25
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Di-tert-butyl dicarbonate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti
EINECSNumber 246-240-1
RTECSNumber HT0230000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.