Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-tert-butyl disulfide

Catalog No. 247545500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. 24-754-5500ML

Supplier: Sigma Aldrich 247545500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mechanism of the oxidation of di-tert-butyl disulfide in the presence of H2O2 catalyzed by bis(acetylacetonato)oxovanadium and a chiral Schiff-base ligand has been investigated.Di-tert-butyl disulfide has been used in the preparation of gold-supported thin film from symmetrical and mixed disulphide adsorbates.

Specifications

CAS 110-06-5
Density 0.923 g/mL (at 25°C (literature))
Boiling Point 200°C to 201°C (lit.)
Molecular Formula C8H18S2
Refractive Index n20/D 1.490 (literature)
Linear Formula (CH3)3 CSSC(CH3)3
MDL Number MFCD00008838
Quantity 500 mL
Synonym tert-Butyl disulfide
Molecular Weight (g/mol) 178.36
Percent Purity 97%
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.