Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Di-tert-butyl Disulfide 97.0+%, TCI America™

Click to view available options
Quantity:
25 mL
100 mL
500 mL
Specifications
Specifications
Color | Yellow |
Boiling Point | 200°C |
Quantity | 25 mL |
UN Number | 3082 |
Formula Weight | 178.35 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Di-tert-butyl Disulfide |
Safety and Handling
EINECSNumber | (2)-0477 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction