Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl hydrazodiformate, 97%

Catalog No. ac440420100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

CAS: 16466-61-8 | C10H20N2O4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16466-61-8
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
InChI Key TYSZETYVESRFNT-UHFFFAOYSA-N
Synonym di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
PubChem CID 85431
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate
SMILES CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C

Specifications

Melting Point 120.0°C to 125.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 232.28
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Di-tert-butyl hydrazodiformate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 240-512-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.