Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl N,N-diethylphosphoramidite, 94%

Catalog No. AAH2738003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2738003 1 g
AAH2738006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2738003 Supplier Thermo Scientific Chemicals Supplier No. H2738003
Add to Cart

CAS: 117924-33-1 | C12H28NO2P | 249.335 g/mol

A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

Solubility
Hydrolyzes in water. Soluble in chloroform and methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117924-33-1
Molecular Formula C12H28NO2P
Molecular Weight (g/mol) 249.335
MDL Number MFCD00134307
InChI Key KUKSUQKELVOKBH-UHFFFAOYSA-N
Synonym di-tert-butyl n,n-diethylphosphoramidite, di-tert-butyl diethylphosphoramidite, di-t-butyl n,n-diethylphosphoramidite, phosphoramidous acid, diethyl-, bis 1,1-dimethylethyl ester, n-bis 2-methylpropan-2-yl oxy phosphanyl-n-ethylethanamine, acmc-1c42i, di-t-butyl diethylphosphoramidite, di-tert-butyl diethylamidophosphite, di-tert-butoxy diethylamino phosphine
PubChem CID 4251849
IUPAC Name N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-ethylethanamine
SMILES CCN(CC)P(OC(C)(C)C)OC(C)(C)C

Specifications

Density 0.896
Boiling Point 39°C to 41°C (0.3 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.434
Quantity 1 g
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Hydrolyzes in water. Soluble in chloroform and methanol.
Formula Weight 249.33
Percent Purity 94%
Chemical Name or Material Di-tert-butyl N,N-diethylphosphoramidite
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.