Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butylchlorophosphine, 96%, AcroSeal™

Catalog No. ac427280500
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle

Catalog No. AC427280500

Supplier: Thermo Scientific Chemicals 427280500

Only null left
Add to Cart

CAS: 13716-10-4 | C8H18ClP | 180.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13716-10-4
Molecular Formula C8H18ClP
Molecular Weight (g/mol) 180.66
MDL Number MFCD00008815
InChI Key MCRSZLVSRGTMIH-UHFFFAOYSA-N
Synonym di-tert-butylchlorophosphine, di-tert-butylchlorophosphane, di-t-butylchlorophosphine, phosphinous chloride, bis 1,1-dimethylethyl, chlorodi-tert-butylphosphine, phosphinous chloride, di-tert-butyl, di-tert-butyl chloro phosphane, di-tert-butylphosphinous chloride, clptbu2, pubchem6477
PubChem CID 139566
IUPAC Name ditert-butyl(chloro)phosphane
SMILES CC(C)(C)P(C(C)(C)C)Cl

Specifications

Melting Point 2.0°C to 3.0°C
Color Colorless
Density 0.9500g/mL
Boiling Point 69.0°C to 70.0°C (10.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula [(CH3)3C]2PCl
Quantity 50 mL
Specific Gravity 0.95
Formula Weight 180.66
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Di-tert-butylchlorophosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.