Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-Butylsilane 95.0+%, TCI America™
SDP

Catalog No. p-7127884
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 30736-07-3
Molecular Formula C8H20Si
MDL Number MFCD00270996

Specifications

Melting Point -38°C
Color Yellow
Boiling Point 129°C
Quantity 1 g
UN Number 1993
Formula Weight 144.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-Butylsilane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.