Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIABLO Monoclonal Antibody (78-1-118)
GREENER_CHOICE

Catalog No. AMPA6846
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6846 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6846 Supplier Invitrogen™ Supplier No. MA116846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: 293 whole cell lysate.

DIABLO (smac), a mitochondrial protein, activates various forms of apoptosis. This activation may be due to the neutralization of one or more members of the IAP family of apoptosis inhibitory proteins. Smac exits the mitochondria and enter the cytosol during certain apoptosis triggered events. Mitochondrial-mediated apoptosis is important in animal development and tissue homeostasis, with alterations resulting in a range of malignant disorders. Upon apoptotic stimuli, the mitochondrial proteins cytochrome c and Smac/DIABLO are released into the cytosol. The release of these proteins, however, occurs via different mechanisms. Smac/DIABLO has also been found to play a key role in regulating the sensitization of cancer cells to apoptosis (both immune and drug-induced).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIABLO
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 78-1-118
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene DIABLO
Gene Accession No. Q9JIQ3, Q9NR28
Gene Alias 0610041G12Rik; 1700006L01Rik; AU040403; DFNA64; DIABLO; diablo homolog (Drosophila); diablo homolog, mitochondrial; diablo IAP-binding mitochondrial protein; diablo, IAP-binding mitochondrial protein; diablo-like protein; DIABLO-S; direct IAP binding protein with low PI; direct IAP-binding protein with low pI; FLJ10537; FLJ25049; hCG_1782202; Second mitochondria-derived activator of caspase; Smac; Smac protein; SMAC3
Gene Symbols DIABLO
Host Species Mouse
Immunogen Recombinant mature human Smac (lacks amino acids 1-55 present in Smac precursor).
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288753, 56616, 66593
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.