Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIABLO Recombinant Rabbit Monoclonal Antibody (006)
DIABLO Recombinant Rabbit Monoclonal Antibody (6)
Supplier: Thermo Scientific MA529220
Description
DIABLO Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA

Specifications
DIABLO | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7 | |
Q9NR28 | |
DIABLO | |
Recombinant Human SMAC/Diablo protein (Ala56-Asp239). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
006 | |
Unconjugated | |
DIABLO | |
0610041G12Rik; 1700006L01Rik; AU040403; DFNA64; DIABLO; diablo homolog (Drosophila); diablo homolog, mitochondrial; diablo IAP-binding mitochondrial protein; diablo, IAP-binding mitochondrial protein; diablo-like protein; DIABLO-S; direct IAP binding protein with low PI; direct IAP-binding protein with low pI; FLJ10537; FLJ25049; hCG_1782202; Second mitochondria-derived activator of caspase; Smac; Smac protein; SMAC3 | |
Rabbit | |
Protein A | |
RUO | |
56616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction