Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetin, 50%, tech. grade, remainder triacetin and monoacetin

Catalog No. p-100654
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle

CAS: 25395-31-7 | C7H12O5 | 176.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetin
Name Note pract., 98%
CAS 25395-31-7, 102-76-1, 26446-35-5
Color Colorless
Density 1.1875g/mL
Boiling Point 280.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 45% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12O5
Refractive Index 1.4400 to 1.4500
Linear Formula (CH3COO)2C3H5OH
Quantity 5 g
MDL Number MFCD00008717
Saponification Value 542 to 605mg KOH/g
Specific Gravity 1.1875
Merck Index 15, 2964
Solubility Information Solubility in water: soluble
Molecular Weight (g/mol) 176.17
Formula Weight 176.17
Percent Purity 50%
Concentration or Composition (by Analyte or Components) 24 to 28% monoacetin, 41 to 47% diacetin, 22 to 28% triacetin (typically)
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.