Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetoxydimethylsilane, 95%, AcroSeal™, Thermo Scientific™

Catalog No. ac434011000
Click to view available options
Quantity:
100 mL

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2182-66-3
Molecular Formula C6H12O4Si
Molecular Weight (g/mol) 176.24
MDL Number MFCD00039832
InChI Key RQVFGTYFBUVGOP-UHFFFAOYSA-N
Synonym dimethyldiacetoxysilane, diacetoxydimethylsilane, dimethylsilanediyl diacetate, silanediol, dimethyl-, diacetate, dimethylsilanediol, diacetate, silanediol, 1,1-dimethyl-, 1,1-diacetate, acetyloxy dimethylsilyl acetate, dimethyldiacetoxysilicane, dimethyl-silanediodiacetate, acmc-1ak12
PubChem CID 75130
IUPAC Name [acetyloxy(dimethyl)silyl] acetate
SMILES CC(=O)O[Si](C)(C)OC(=O)C

Specifications

Melting Point -12.5°C
Density 1.0530g/mL
Boiling Point 164°C
Flash Point 69°C
Assay Percent Range 94% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3COO)2Si(CH3)2
Quantity 100 mL
Specific Gravity 1.053
Formula Weight 176.24
Percent Purity 95%
Chemical Name or Material Diacetoxydimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
EINECSNumber 218-562-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.