Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetyllauroyl Glycerol (so called) [Plasticizer], TCI America™
SDP

Catalog No. D2482500ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

MDL Number MFCD00671531

Specifications

Color Yellow
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Diacetyllauroyl Glycerol (so called) [Plasticizer]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.