Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide

Catalog No. aal1110614
Click to view available options
Quantity:
25 g
100 g

Diallyl disulfide, 80%, C6H10S2, CAS Number-2179-57-9, diallyldisulfide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene, 2-propenyl disulphide, diallyl disulfide, allyl disulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 100g | CAS: 2179-57-9 | C6H10S2 | 146.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2179-57-9
Molecular Formula C6H10S2
Molecular Weight (g/mol) 146.27
MDL Number MFCD00008656
InChI Key PFRGXCVKLLPLIP-UHFFFAOYSA-N
Synonym diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
PubChem CID 16590
ChEBI CHEBI:4488
IUPAC Name 3-(prop-2-enyldisulfanyl)prop-1-ene
SMILES C=CCSSCC=C

Specifications

Density 1.008
Boiling Point 180°C to 190°C
Flash Point 51°C (123°F)
Odor Garlic-like
Refractive Index 1.541
Linear Formula CH2=CHCH2S-SCH2CH=CH2
Quantity 25 g
UN Number UN1993
Beilstein 1699241
Formula Weight 146.27
Percent Purity 80%
Grade Technical
Assay remainder mainly diallyl sulfide and diallyl trisulfide
Chemical Name or Material Diallyl disulfide
Show More Show Less
Hazard Category H226-H301-H315-H317-H319
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 218-548-6
RTECSNumber BB1000000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.