Learn More
Diamedix™ Sample Racks for MAGO™ Plus and MAGO™ 4S Automated Immunoassay System
SDP
Catalog No. 23451069
Encompass
Click to view available options
Type:
Sample Rack A F/13X100
Sample Rack A F/13X75
Sample Rack A F/16X100
Sample Rack B F/13X100
Sample Rack B F/13X75
Sample Rack B F/16X100
Accessory for use with MAGO Plus and MAGO 4S automated immunoassay systems.
For use with MAGO Plus and MAGO 4S automated immunoassay systems.
Specifications
Type Sample Rack B F/13X75
For Use With (Application) ELISA
For Use With (Equipment) 13 x 75mm Primary tubes; for MAGO™ Plus and MAGO™ 4S automated immunoassay system
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.