Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIAPH-2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600280H
Description
DIAPH-2 Polyclonal antibody specifically detects DIAPH-2 in Human samples. It is validated for Western BlotSpecifications
DIAPH-2 | |
Polyclonal | |
Western Blot | |
DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2 | |
The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Diaphanous Homolog 2 using the numbering given in entry NP_006720.1 (GeneID 1730). | |
0.1 mL | |
Cell Cycle and Replication, Transcription Factors and Regulators | |
1730 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction