Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIAPH-2 Antibody (V78P3C10*D3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250229AF594
Description
DIAPH-2 Monoclonal specifically detects DIAPH-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
DIAPH-2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
DIAPH2 | |
Peptide Sequence - RHNGAISSK | |
0.1 mL | |
1730 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry | |
V78P3C10*D3 | |
Western Blot, ELISA, Immunohistochemistry | |
DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction