Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIAPH-2 Antibody (V78P3C10*D3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250229MFV500
Description
DIAPH-2 Monoclonal antibody specifically detects DIAPH-2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| DIAPH-2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| V78P3C10*D3 | |
| Western Blot, ELISA, Immunohistochemistry | |
| DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2 | |
| Peptide Sequence - RHNGAISSK | |
| 0.1 mL | |
| Cell Cycle and Replication, Transcription Factors and Regulators | |
| 1730 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction