Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIAPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596420 Supplier Invitrogen™ Supplier No. PA596420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEEKMRRAKL AKEKAEKERL EKQQKREQLI DMNAEGDETG VMDSLLEALQ SGAAFRRKRG PRQANRKAGC AVTSLLASEL TKDDAMAAVP AKVSKNSETF PTILEEAKEL VGRAS; Positive Samples: HL-60, MCF7, 293T, HepG2, Mouse liver, Mouse lung; Cellular Location: Cell membrane, Cell projection, Cytoplasm, cytoskeleton, ruffle membrane.

This gene is a homolog of the Drosophila diaphanous gene, and has been linked to autosomal dominant, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. It has therefore been speculated that this gene may have a role in the regulation of actin polymerization in hair cells of the inner ear. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIAPH1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DIAPH1
Gene Accession No. O08808, O60610
Gene Alias D18Wsu154e; DFNA1; DIA1; Diap1; Diaph1; diaphanous 1; diaphanous homolog 1; diaphanous homolog 1 (Drosophila); diaphanous related formin 1; diaphanous-1; diaphanous-related formin 1; Diaphanous-related formin-1; DRF1; FLJ25265; hDIA1; LFHL1; mDIA1; p140DIA; p140mDia; Protein diaphanous homolog 1; SCBMS
Gene Symbols DIAPH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1149-1263 of human DIAPH1 (NP_0010732801).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13367, 1729
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.