Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIAPH1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA519033
Description
This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene is a homolog of the Drosophila diaphanous gene, and has been linked to autosomal dominant, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. It has therefore been speculated that this gene may have a role in the regulation of actin polymerization in hair cells of the inner ear. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| DIAPH1 | |
| Polyclonal | |
| Unconjugated | |
| DIAPH1 | |
| D18Wsu154e; DFNA1; DIA1; Diap1; Diaph1; diaphanous 1; diaphanous homolog 1; diaphanous homolog 1 (Drosophila); diaphanous related formin 1; diaphanous-1; diaphanous-related formin 1; Diaphanous-related formin-1; DRF1; FLJ25265; hDIA1; LFHL1; mDIA1; p140DIA; p140mDia; Protein diaphanous homolog 1; SCBMS | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 13367 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O08808 | |
| DIAPH1 | |
| Synthetic peptide sequence (QTKTSKAKKDQ) corresponding to the internal amino acids of DIAPH1 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction