Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIAPH1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303284100UL
Description
DIAPH1 Polyclonal specifically detects DIAPH1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
DIAPH1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
DIA1, diaphanous homolog 1 (Drosophila), hDIA1, homolog) 1, LFHL1FLJ25265 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1149-1263 of human DIAPH1 (NP_001073280.1). | |
100 μL | |
Signal Transduction | |
1729 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction