Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIAPH2/Dia2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982533
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2533 Supplier Bioss Supplier No. BS13002R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The product of this gene belongs to the diaphanous subfamily of the formin homology family of proteins. This gene may play a role in the development and normal function of the ovaries. Defects in this gene have been linked to premature ovarian failure 2. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen DIAPH2/Dia2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DIAPH2
Gene Accession No. O60879
Gene Alias DIA; DIA2; Dia3; Diap2; Diaph2; diaphanous homolog 2; diaphanous homolog 2 (Drosophila); diaphanous related formin 2; diaphanous-related formin 2; Diaphanous-related formin-2; diaphorase-2; DRF2; E430022I22Rik; FLJ11167; mDia3; mDia3 isoform; POF; POF2; Protein diaphanous homolog 2; RGD1564399; RP11-212B22.1
Gene Symbols DIAPH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DIAPH2/Dia2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1730
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.