Learn More
Invitrogen™ DIAPH2 Monoclonal Antibody (CL1111)

Description
Immunogen sequence: GGSEEPGGGR SNKRSAGNRA ANEEETKNKP KLNIQIKTLA DDVRDRITSF RKSTVKKEKP LIQHPIDSQV AMSEFPAAQP LYDERSLNLS EKEVLDLFEK MMEDMNLNEE KKAPLRNKDF TTKREMVVQY ISAT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 33%. Binds to an epitope located within the peptide sequence SLNLSEKEVLDLFEK as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | DIAPH2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL1111 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DIAPH2 |
| Gene Accession No. | O60879 |
| Gene Alias | DIA; DIA2; Dia3; Diap2; Diaph2; diaphanous homolog 2; diaphanous homolog 2 (Drosophila); diaphanous related formin 2; diaphanous-related formin 2; diaphanous-related formin-2; diaphorase-2; DRF2; E430022I22Rik; FLJ11167; mDia3; mDia3 isoform; POF; POF2; Protein diaphanous homolog 2; RGD1564399; RP11-212B22.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.