Learn More
Invitrogen™ DIAPH2 Polyclonal Antibody

Description
PA1-31027 detects DIAPH2 from human samples.
Specifications
Specifications
| Antigen | DIAPH2 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.15M NaCl and 0.01% sodium azide; pH 7.2 |
| Gene | DIAPH2 |
| Gene Accession No. | O60879 |
| Gene Alias | DIA; DIA2; Dia3; Diap2; Diaph2; diaphanous homolog 2; diaphanous homolog 2 (Drosophila); diaphanous related formin 2; diaphanous-related formin 2; diaphanous-related formin-2; diaphorase-2; DRF2; E430022I22Rik; FLJ11167; mDia3; mDia3 isoform; POF; POF2; Protein diaphanous homolog 2; RGD1564399; RP11-212B22.1 |
| Gene Symbols | DIAPH2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.