Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIAPH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115123 Supplier Invitrogen™ Supplier No. PA5115123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DIAPH3.

This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal dominant auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIAPH3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DIAPH3
Gene Accession No. Q9NSV4, Q9Z207
Gene Alias 4930417P13Rik; AN; AUNA1; DIA2; diap3; Diaph3; diaphanous homolog 3; diaphanous homolog 3 (Drosophila); diaphanous related formin 3; diaphanous-related formin; diaphanous-related formin 3; diaphanous-related formin-3; DKFZp434C0931; DKFZp686A13178; Drf3; FLJ34705; mDia2; mKIAA4117; NSDAN; p134MDia2; Protein diaphanous homolog 3
Gene Symbols DIAPH3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DIAPH3(Accession Q9NSV4), corresponding to amino acid residues T825-S875.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290396, 56419, 81624
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.