Learn More
Diaphanous 1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Diaphanous 1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2326
![]() |
Bethyl Laboratories, Inc
IHC00571T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2325
![]() |
Bethyl Laboratories, Inc
IHC00571 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla and Northern white-cheeked gibbon
This gene is a homolog of the Drosophila diaphanous gene, and has been linked to autosomal domit, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. It has therefore been speculated that this gene may have a role in the regulation of actin polymerization in hair cells of the inner ear. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
Diaphanous 1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DIAPH1; diaphanous homolog 1 (Drosophila); DFNA1; DIA1; DRF1; FLJ25265; LFHL1; hDIA1; diaphanous 1; diaphanous-1; diaphanous-related formin 1 | |
DIAPH1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O60610 | |
1729 | |
Between 1200 and C-term | |
Primary | |
4° C | |
DIAPH1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.