Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatomaceous Earth, Spectrum™ Chemical
SDP

Catalog No. 18602134 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Pail

DI106, 68855-54-9

Spectrum™ Chemical Diatomaceous Earth is also known as diatomite and occurs naturally as a soft sedimentary rock that is easily crumbled into a white colored powder. It can have an abrasive texture similar to that of pumice powder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Quantity 12 kg
CAS 68855-54-9
CAS Max % 100%
Packaging Fiber Drum
Grade Reagent
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.