Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diazo Biotin-Alkyne

Catalog No. 90199525MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
100 mg

Catalog No. 90-199-525MG

Supplier: Sigma Aldrich 90199525MG

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diazo biotin-alkyne is an alkyne-activated, sodium dithionite (Na2S2O4) cleavable biotin probe that allows for efficient release of the captured biomolecules from streptavidin. This reagent contains a biotin moiety linked to an azide group through a spacer arm containing a diazo group that can be cleaved with 50 mM sodium dithionite solution.

Specifications

CAS 142356-33-0
Molecular Formula C22H25NO5 -+ HCl
Linear Formula C22H25NO5 -+ HCl
Quantity 100 mg
Synonym Amino (PEG)3 benzophenone alkyne HCl salt; Probe building block
Molecular Weight (g/mol) 419.9 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.