Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibasic Potassium Phosphate, Anhydrous, EP, BP, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602147 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-602-147

Supplier: Spectrum Chemical Manufacturing Corporation PO20450KGBL

Only null left
Add to Cart
Add to Cart

PO204, 7758-11-4, K2HPO4

Spectrum™ Chemical Dibasic Potassium Phosphate, Anhydrous, USP, EP, BP is used as a mineral supplement and is categorized in the United States by the Food and Drug Administration as generally recognized as safe (GRAS). Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

pH 8.5-9.6
CAS Max % 100%
Quantity 50 kg
Percent Purity 98 to 100.5%
Grade Multi-Compendial
Loss on Drying 1%
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.