Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo[a,e]cyclooctene 97.0+%, TCI America™
SDP

Catalog No. D43381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43381G 1 g
D4338200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43381G Supplier TCI America Supplier No. D43381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 262-89-5
Molecular Formula C16H12
Molecular Weight (g/mol) 204.272
InChI Key VGQWCQNHAMVTJY-YAZGSDAZSA-N
Synonym Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene, DBCOT
PubChem CID 6439814
IUPAC Name (4aZ,6Z,10aZ,12Z)-dibenzo[2,1-a
SMILES C1=CC2=CC=C3C=CC=CC3=CC=C2C=C1

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 204.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo[a,e]cyclooctene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.