Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzosuberyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D246125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D246125G 25 g
D24615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D246125G Supplier TCI America Supplier No. D246125G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1210-33-9
Molecular Formula C15H13Cl
Molecular Weight (g/mol) 228.72
MDL Number MFCD00003592
InChI Key QPERNSDCEUTOTE-UHFFFAOYSA-N
Synonym 5-chloro-10,11-dihydro-5h-dibenzo a,d cycloheptene, 5-chlorodibenzosuberane, dibenzosuberyl chloride, 5-chloro-10,11-dihydro-5h-dibenzo a,d 7 annulene, 5h-dibenzo a,d cycloheptene, 5-chloro-10,11-dihydro, 5h-dibenzo a,d cycloheptene, 10,11-dihydro-5-chloro, 2-chlorotricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaene, 2-chlorotricyclo 9.4.0.0 3 ,? pentadeca-1 11 ,3,5,7,12,14-hexaene, dibenzosuberylchloride
PubChem CID 14587
IUPAC Name 2-chlorotricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
SMILES ClC1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 25 g
Formula Weight 228.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzosuberyl Chloride
RTECSNumber HO8302000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.