Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene-2-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. D47771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47771G 1 g
D47775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47771G Supplier TCI America Supplier No. D47771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22099-23-6
Molecular Formula C13H8OS
Molecular Weight (g/mol) 212.266
MDL Number MFCD02251670
InChI Key QFGXAEFJJINHAI-UHFFFAOYSA-N
PubChem CID 641360
IUPAC Name dibenzothiophene-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)C=O

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzothiophene-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.