Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene-4-boronic acid, 95%

Catalog No. p-4323515
Click to view available options
Quantity:
1 g
5 g

CAS: 108847-20-7 | C12H9BO2S | 228.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108847-20-7
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.07
MDL Number MFCD01318182
InChI Key GOXNHPQCCUVWRO-UHFFFAOYSA-N
Synonym dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
PubChem CID 2734329
SMILES OB(O)C1=CC=CC2=C1SC1=C2C=CC=C1

Specifications

Melting Point 285.0°C to 290.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name {8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid
Formula Weight 228.08
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Dibenzothiophene-4-boronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.