Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene, 98%

Catalog No. p-7082004
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 132-65-0 | C12H8S | 184.256 g/mol

Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-65-0
Molecular Formula C12H8S
Molecular Weight (g/mol) 184.256
MDL Number MFCD00004969
InChI Key IYYZUPMFVPLQIF-UHFFFAOYSA-N
Synonym diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene
PubChem CID 3023
ChEBI CHEBI:23681
IUPAC Name dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2

Specifications

Melting Point 97°C to 100°C
Boiling Point 332°C to 333°C
Flash Point 170°C (338°F)
Quantity 25 g
UN Number UN3077
Beilstein 121101
Solubility Information Soluble in water.
Formula Weight 184.25
Percent Purity 98%
Chemical Name or Material Dibenzothiophene
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-072-9
RTECSNumber HQ3490550
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.