Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dibenzoyl-L-tartaric anhydride, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
Plastic bottle
64339-95-3
C18H12O7
340.29
MFCD00067102
OXIKRMSPXYQFOT-ZIAGYGMSSA-N
3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl dibenzoate,+-dibenzoyl-l-tartaric anhydride,2,5-furandione, 3,4-bis benzoyloxy dihydro-, 3r,4r,2-o,3-o-dibenzoyl-l-tartaric anhydride,3r,4r-2,5-dioxo-3,4-dibenzoyloxy-3,4-dihydrofuran,3r,4r-4-benzoyloxy-2,5-dioxooxolan-3-yl benzoate
2733743
[(3R,4R)-4-benzoyloxy-2,5-dioxooxolan-3-yl] benzoate
C1=CC=C(C=C1)C(=O)OC2C(C(=O)OC2=O)OC(=O)C3=CC=CC=C3
64339-95-3
C18H12O7
340.29
MFCD00067102
OXIKRMSPXYQFOT-ZIAGYGMSSA-N
3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl dibenzoate,+-dibenzoyl-l-tartaric anhydride,2,5-furandione, 3,4-bis benzoyloxy dihydro-, 3r,4r,2-o,3-o-dibenzoyl-l-tartaric anhydride,3r,4r-2,5-dioxo-3,4-dibenzoyloxy-3,4-dihydrofuran,3r,4r-4-benzoyloxy-2,5-dioxooxolan-3-yl benzoate
2733743
[(3R,4R)-4-benzoyloxy-2,5-dioxooxolan-3-yl] benzoate
C1=CC=C(C=C1)C(=O)OC2C(C(=O)OC2=O)OC(=O)C3=CC=CC=C3
Melting Point 174°C
Quantity 5g
Packaging Glass Bottle
Assay Percent Range 98%
Solubility Information Solubility in water: .. Other solubilities: soluble in ethyl ether
Formula Weight 340.29
Percent Purity 98%
Chemical Name or Material (+)-Dibenzoyl-L-tartaric anhydride

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA