Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl carbonate, 98%

Catalog No. AAA1146418
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 3459-92-5 | C15H14O3 | 242.27 g/mol

Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

Solubility
Sparingly Soluble (0.099 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3459-92-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00014436
InChI Key PIZLBWGMERQCOC-UHFFFAOYSA-N
Synonym carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester
PubChem CID 77002
IUPAC Name dibenzyl carbonate
SMILES O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Melting Point 27°C to 30°C
Boiling Point 150°C to 155°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5485
Quantity 50 g
Beilstein 1882864
Sensitivity Air sensitive
Solubility Information Sparingly Soluble (0.099g/L) (25°C).
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material Dibenzyl carbonate
Show More Show Less
Hazard Category H302+H312
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 222-401-1
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.