Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl Dicarbonate 97.0+%, TCI America™
SDP

Catalog No. P12815G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 31139-36-3
Molecular Formula C16H14O5
Molecular Weight (g/mol) 286.28
MDL Number MFCD00043124
InChI Key FHRRJZZGSJXPRQ-UHFFFAOYSA-N
Synonym Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester
PubChem CID 3649378
IUPAC Name dibenzyl dicarbonate
SMILES O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 286.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl Dicarbonate
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.