Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl diisopropylphosphoramidite, 95%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
108549-23-1
C20H28NO2P
345.423
MFCD00191988
ANPWLBTUUNFQIO-UHFFFAOYSA-N
dibenzyl diisopropylphosphoramidite,dibenzyl n,n-diisopropylphosphoramidite,bis benzyloxy diisopropylamino phosphine,dibenzyldiisopropylphosphoramidite,dibenzyloxy diisopropylamino phosphine,bis benzyloxy phosphanyl diisopropylamine,phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester,n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine,bis-bzl-2ipr-p,bis methylethyl bis phenylmethoxy phosphino amine
196621
N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2
108549-23-1
C20H28NO2P
345.423
MFCD00191988
ANPWLBTUUNFQIO-UHFFFAOYSA-N
dibenzyl diisopropylphosphoramidite,dibenzyl n,n-diisopropylphosphoramidite,bis benzyloxy diisopropylamino phosphine,dibenzyldiisopropylphosphoramidite,dibenzyloxy diisopropylamino phosphine,bis benzyloxy phosphanyl diisopropylamine,phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester,n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine,bis-bzl-2ipr-p,bis methylethyl bis phenylmethoxy phosphino amine
196621
N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2
Applications
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.
Boiling Point 130°C (0.55mmHg)
Quantity 5g
Flash Point 69°C (156°F)
Assay Percent Range 0.9
Formula Weight 345.43
Percent Purity 95%
Chemical Name or Material Dibenzyl diisopropylphosphoramidite

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : Moisture Sensitive

Recommended Storage : 2° to 8°C