Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl diisopropylphosphoramidite, 90+%

Catalog No. AAB2262914
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 108549-23-1 | C20H28NO2P | 345.423 g/mol

Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108549-23-1
Molecular Formula C20H28NO2P
Molecular Weight (g/mol) 345.423
MDL Number MFCD00191988
InChI Key ANPWLBTUUNFQIO-UHFFFAOYSA-N
Synonym dibenzyl diisopropylphosphoramidite, dibenzyl n,n-diisopropylphosphoramidite, bis benzyloxy diisopropylamino phosphine, dibenzyldiisopropylphosphoramidite, dibenzyloxy diisopropylamino phosphine, bis benzyloxy phosphanyl diisopropylamine, phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester, n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine, bis-bzl-2ipr-p, bis methylethyl bis phenylmethoxy phosphino amine
PubChem CID 196621
IUPAC Name N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2

Specifications

Density 1.03
Boiling Point 130°C (0.55 mmHg)
Flash Point 69°C (156°F)
Assay Percent Range 0.9
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 345.43
Percent Purity 95%
Chemical Name or Material Dibenzyl diisopropylphosphoramidite
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 629-628-9
TSCA No
storageNote1 Moisture Sensitive
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.