Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzylamine, 98%

Catalog No. AC112610010
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112610010 1 L Glass bottle
AC112612500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112610010 Supplier Thermo Scientific Chemicals Supplier No. 112610010
Add to Cart

CAS: 103-49-1 | C14H15N | 197.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-49-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00004770
InChI Key BWLUMTFWVZZZND-UHFFFAOYSA-N
Synonym dibenzylamine, benzenemethanamine, n-phenylmethyl, n-benzylbenzylamine, bibenzylamine, dibenzyl-amine, dibenzyl amine, bisbenzylamine, n,n-dibenzylamine, n-benzylaminomethyl benzene, unii-3g0yfx01c6
PubChem CID 7656
SMILES C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -26°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 300°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5740 to 1.576
Linear Formula (C6H5CH2)2NH
Quantity 1 L
Beilstein 12, 1035
Specific Gravity 1.02
Merck Index 15, 3016
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol,ether,toluene and chloroform,reacts with organic and inorganic acids
IUPAC Name N-benzyl-1-phenylmethanamine
Formula Weight 197.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dibenzylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/f
EINECSNumber 203-117-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.