Learn More
Dibromo-tyrosine Antibody (9F12), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259351C
Catalog No. NBP259351C
Only null left
Add to Cart
Description
Description
Dibromo-tyrosine Monoclonal specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Dibromo-tyrosine
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine.
Store at 4C in the dark.
IgG1
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
9F12
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein G purified
Primary
Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.