Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibromo-tyrosine Antibody (9F12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259351H
Description
Dibromo-tyrosine Monoclonal specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA.Specifications
Dibromo-tyrosine | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
9F12 | |
Western Blot, ELISA | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction