Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibromo-tyrosine, Mouse anti-Human, Clone: 6G3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16480
Specifications
Dibromo-tyrosine | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
RUO | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
6G3 | |
ELISA (1:1000) Immunocytochemistry (1:50) Immunofluorescence (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction