Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromobis(triphenylphosphine)nickel(II), 99%, Thermo Scientific Chemicals

Catalog No. AA4219206
Click to view available options
Quantity:
1 g
5 g

As a polymerization catalyst

As a polymerization catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14126-37-5
Molecular Formula C36H30Br2NiP2
Molecular Weight (g/mol) 743.09
MDL Number MFCD00015865
InChI Key QEKXARSPUFVXIX-UHFFFAOYSA-L
Synonym nickel ii bromide bis triphenylphosphine, nibr2 pph3 2, dibromobis triphenylphosphine nickel ii, bis triphenylphosphine nickel ii bromide, dibromobis triphenylphosphine nickel, dibromonickel; bis triphenylphosphine, bis triphenylphosphine dibromonickel ii
PubChem CID 11050914
IUPAC Name dibromonickel;triphenylphosphane
SMILES Br[Ni]Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 219°C to 223°C
Odor Odorless
Quantity 5 g
Formula Weight 743.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)
Hazard Category H302+H312+H332-H314-H350
Hazard Statement GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.