Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibromochloromethane, 97+%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It affords bromochlorocarbene under phase-transfer conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It affords bromochlorocarbene under phase-transfer conditions.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
It affords bromochlorocarbene under phase-transfer conditions.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
Specifications
Specifications
| Melting Point | -22°C |
| Density | 2.42 |
| Boiling Point | 115°C to 118°C |
| Refractive Index | 1.5475 |
| Quantity | 5 g |
| Beilstein | 1731046 |
| Sensitivity | Light and moisture sensitive |
| Merck Index | 14,2135 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 208.28 |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H341-H302 Suspected of causing genetic defects. Harmful if swallowed. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 204-704-0 |
| RTECSNumber | PA6360000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction