Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyl disulfide

Catalog No. B93989100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B93989100ML 100 mL
B9398925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B93989100ML Supplier Sigma Aldrich Supplier No. B93989100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutyl disulfide

Specifications

CAS 629-45-8
Density 0.938 g/mL (at 25°C (literature))
Boiling Point 229°C to 233°C (lit.)
Molecular Formula C8H18S2
Refractive Index n20/D 1.492 (literature)
Linear Formula CH3(CH2)3 SS(CH2)3 CH3
Quantity 100 mL
MDL Number MFCD00009467
Synonym Butyl disulfide
Molecular Weight (g/mol) 178.36
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.