Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl L-tartrate 98%, Thermo Scientific™

Catalog No. AC406660050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87-92-3
Density 1.0900g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C12H22O6
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Linear Formula CH3(CH2)3OCOCHOHCHOHCOO(CH2)3CH3
Quantity 5g
MDL Number MFCD00009443
Specific Rotation Condition + 10.50 (20.00°C c=neat)
Specific Rotation +10° to +11° (20°C, 589nm) (neat)
Synonym 2r,3r-dibutyl 2,3-dihydroxysuccinate, dibutyl l-+-tartrate, dibutyl l-tartrate, unii-2d1v32if1e, dibutyl tartrate, +, l-+-tartaric acid di-n-butyl ester, dibutyl 2r,3r-2,3-dihydroxybutanedioate, butanedioic acid, 2,3-dihydroxy-2r,3r-, dibutyl ester, 1,4-dibutyl 2r,3r-2,3-dihydroxybutanedioate, dibutyll-+-tartrate
InChI Key PCYQQSKDZQTOQG-NXEZZACHSA-N
SMILES CCCCOC(=O)C(C(C(=O)OCCCC)O)O
IUPAC Name dibutyl (2R,3R)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol) 262.302
PubChem CID 6910
Formula Weight 262.29
Percent Purity ≥97.5%
Chemical Name or Material Dibutyl L-tartrate, 98%
Show More Show Less
EINECSNumber 201-784-9
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.