Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibutyl Methylenebis(thioglycolate) 95.0+%, TCI America™

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Color | Colorless |
Boiling Point | 178°C |
Quantity | 25 g |
Formula Weight | 308.45 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | Dibutyl Methylenebis(thioglycolate) |
Safety and Handling
EINECSNumber | (2)-1401 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction