Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl phosphite, 14.5-16% P, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
1809-19-4
C8H19O3P
194.21
MFCD00066633
OSPSWZSRKYCQPF-UHFFFAOYSA-N
dibutyl phosphite,dibutyl phosphonate,phosphonic acid, dibutyl ester,di-n-butylphosphite,dibutoxyphosphine oxide,mobil dbhp,dibutyl hydrogen phosphonate,di-n-butyl hydrogen phosphite,dibutylfosfit,phosphorous acid, dibutyl ester
6327349
dibutoxy(oxo)phosphanium
CCCCO[P+](=O)OCCCC
1809-19-4
C8H19O3P
194.21
MFCD00066633
OSPSWZSRKYCQPF-UHFFFAOYSA-N
dibutyl phosphite,dibutyl phosphonate,phosphonic acid, dibutyl ester,di-n-butylphosphite,dibutoxyphosphine oxide,mobil dbhp,dibutyl hydrogen phosphonate,di-n-butyl hydrogen phosphite,dibutylfosfit,phosphorous acid, dibutyl ester
6327349
dibutoxy(oxo)phosphanium
CCCCO[P+](=O)OCCCC
Infrared Spectrum Authentic
Quantity 500g
Density 0.9950g/mL
Boiling Point 118°C to 119°C (11.0mmHg)
Specific Gravity 0.995
Flash Point 121°C
Refractive Index 1.4225 to 1.4245
Packaging Glass bottle
Color Undesignated
Assay Percent Range 96%
Linear Formula [CH3(CH2)3O]2P(O)H
Beilstein 01, I, 187
Solubility Information Solubility in water: slightly soluble
Formula Weight 194.21
Physical Form Liquid
Percent Purity 14.5 to 16% (P)
Chemical Name or Material Dibutyl phosphite, 96%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-316-1

RTECSNumber : HS6475000

TSCA : TSCA